Learn More
Thermo Scientific™ Chromogenic Listeria Agar (ISO)
Description
Isolate and detect Listeria monocytogenes from food samples using Thermo Scientific™ Chromogenic Listeria Agar (ISO), a specially formulated media which conforms to ISO 11290-1 and 11290-2. The expression of the phospholipase C enzyme produced by L. monocytogenes and Listeria ivanovii creates a unique opaque white halo around the L. mono colony which is the diagnostic criteria necessary to differentiate from non-pathogenic Listeria spp.
This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | According ISO 11290-1 and ISO 11290-2 |
| Color | Straw, semi-opaque |
| Content And Storage | 2°C to 10°C |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Media Type | CLA (ISO) |
| Product Type | Agar |
| Fill Weight | 19.5g ± 1.0g |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.