missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromofil™ Polyester Syringe Filter

Filter polar or nonpolar solvents. Chromofil™ PET Polyester Syringe Filter is hydrophilic for multipurpose applications and comes color coded.

Brand:  Macherey-Nagel™ 729023

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 10209411

  • £111.00 / Pack of 100
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

  • Syringe filters with polyester membrane  
  • Color-coded 
  • Hydrophilic multipurpose membrane
  • For polar as well as nonpolar solvents
  • The HPLC filter, especially suited for mixtures of water and organic solvents 
  • For TOC/DOC determination
  • Not cytotoxic, does not inhibit the growth of microorganisms and higher cells
Specifications

Specifications

Colorless Top and Orange Bottom
0.45 μm
Hydrophilic
Syringe Filter
Polyester
Non Polar and Polar Solvent
100 Pack
15 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers