Learn More
Chromium boride, 99% (metals basis), Thermo Scientific Chemicals
£57.30 - £166.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337417
|
Thermo Scientific Alfa Aesar
012570.14 |
25 g |
£57.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347417
|
Thermo Scientific Alfa Aesar
012570.22 |
100 g |
£166.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromium boride,is been used in plasma technology and for high-performance electrodes in electron microscopes, metal matrix composites (MMCs), cutting tools and cermets for machining aluminum. chromium diboride is recognised as offering superior performance in corrosive-wear environments. Chromium diboride thin films possess desirable combinations of properties, which are attractive for a wide range of potential industrial applications. As a wear-resistant component of weld materials and powders for spraying in engineering and metallurgy, Protection of outer surfaces and bearing surfaces of machine parts against abrasive wear by plasma spraying, In aluminum evaporators manufacturing and for aluminum electrolyzes repair work, In nuclear engineering as neutron absorption material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12007-16-8 | |
CrB2 | |
MFCD00016043 |
Specifications
12007-16-8 | |
Powder | |
CrB2 | |
It is insoluble in water. | |
Odorless |
Gray | |
99% (Metals basis) | |
MFCD00016043 | |
73.62 | |
Chromium boride |
Safety and Handling
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i
missing translation for 'einecsNumber' : 234-499-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only