missing translation for 'onlineSavingsMsg'
Learn More

Chromium boride, 99% (metals basis)

Product Code. 11347417
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11347417

Brand: Thermo Scientific Alfa Aesar 012570.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12007-16-8 | CrB2

Chromium boride,is been used in plasma technology and for high-performance electrodes in electron microscopes, metal matrix composites (MMCs), cutting tools and cermets for machining aluminum. chromium diboride is recognised as offering superior performance in corrosive-wear environments. Chromium diboride thin films possess desirable combinations of properties, which are attractive for a wide range of potential industrial applications. As a wear-resistant component of weld materials and powders for spraying in engineering and metallurgy, Protection of outer surfaces and bearing surfaces of machine parts against abrasive wear by plasma spraying, In aluminum evaporators manufacturing and for aluminum electrolyzes repair work, In nuclear engineering as neutron absorption material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-16-8
Molecular Formula CrB2
MDL Number MFCD00016043

Specifications

Color Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% (Metals basis)
Solubility Information It is insoluble in water.
Formula Weight 73.62
Odor Odorless
Chemical Name or Material Chromium boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.