missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ CHROMagar™ O157 Medium (powder)

Product Code. 10111253 Shop All Applied Biosystems Products
Click to view available options
:
4 x 250mL
This item is not returnable. View return policy

Product Code. 10111253

Brand: Applied Biosystems™ 74002

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMagar™ O157 Medium

Recommended as a second media to CT-SMAC. CHROMagar™ distinguishes between Pseudomonas spp and E.coli O157 by a specific colour reaction.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate, store at 2°C to 8°C
Form Powder
Media Type Pathogen Detection Media
Preparation Method Microwave
Product Type Agar
Target Organism Class E. coli
Final Product Type Agar Plates
Product Line CHROMagar
Shipping Condition Room Temperature
Strain Designations E. coli O157

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.