Learn More
Macherey-Nagel™ Chromafix™ SiOH Cartridge
Product Code. 10063191 Shop All Macherey-Nagel Products
Change view
Click to view available options
Description:
Adsorbent Weight: 230 mg
Adsorbent Weight: 420 mg
Adsorbent Weight: 880 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10063191 Adsorbent Weight: 230 mg Pack of 50
10411862 Adsorbent Weight: 420 mg Pack of 50
10023031 Adsorbent Weight: 880 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10063191 Supplier Macherey-Nagel™ Supplier No. 731828
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Base Material Weakly Acidic Silica
Description Adsorbent Weight: 230 mg
For Use With (Application) Aflatoxins, Chloramphenicol, Pesticides, Steroids, Vitamins
Particle Size 45 μm
Sorbent SiOH
Type Chromafix SiOH Cartridge
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g