Learn More
Macherey-Nagel™ Chromafix™ SiOH Cartridge
Brand: Macherey-Nagel™ 731829
Code : 50
Additional Details : Weight : 0.01000kg
Specifications
Weakly Acidic Silica | |
Aflatoxins, Chloramphenicol, Pesticides, Steroids, Vitamins | |
SiOH | |
60 Å | |
500 m2/g |
Adsorbent Weight: 420 mg | |
45 μm | |
Chromafix SiOH Cartridge | |
50 Pack |