Learn More
Description
Polypropylene SPE columns with a base material of coarse-grained kieselguhr (also known as diatomaceous earth, hydromatrix, celite), with a large pore size and high pore volume. The pH value of aqueous solutions can be altered on the column, from 1–13, which enables elution of different compounds of a sample under optimized conditions.
Application:
For liquid-liquid extraction of highly viscous aqueous solutions without use of a separation funnel. Columns feature high water loadability without breakthrough during elution with organic solvents. They are also suited for removing small amounts of water from solvents which are not miscible with water.
- Physiological fluids (blood, plasma, and serum) in clinical chemistry
- Dyes in textiles
- Environmental and food analysis
Advantages:
- Fast, reproducible and economical
- Simultaneous preparation of several samples
- No problems with phase separation—no formation of emulsions
- High recovery rates
- Saving of time and solvents
- Organic solutions do not need to be dried after separation
Specifications
Specifications
| Column Capacity (Metric) | 45 mL |
| Column Type | Polypropylene |
| Bed Weight | 8.3 g |
| For Use With (Application) | Liquid-Liquid Extraction |
| Sorbent | Chromabond XTR |
| Type | Chromabond XTR Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.