Learn More
Macherey-Nagel™ CHROMABOND™ Complete Vacuum Manifold
Product Code. 10236331 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
up to 12 Columns
up to 16 LV Columns
up to 24 Columns
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10236331 up to 16 LV Columns Each
10799172 up to 12 Columns Each
10025731 up to 24 Columns Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10236331 Supplier Macherey-Nagel™ Supplier No. 730360
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for the simultaneous preparation of up to 12, 16 or 24 samples. Macherey-Nagel™ CHROMABOND™ Complete Vacuum Manifold handles columns and cartridges used in solid phase extraction (SPE).
The complete CHROMABOND Vacuum Manifold consists of:
  • Rectangular glass cabinet with polypropylene lid and lid gasket (2 sizes available):
    • Small holds up to 12 CHROMABOND™ columns or CHROMAFIX™ cartridges
    • Large holds up to 16 CHROMABOND™ LV columns or up to 24 CHROMABOND™ columns or CHROMAFIX™ cartridges
  • Removable needles on lower side of the lid
  • Vacuum gauge for reading the vacuum pressure in the chamber
  • Control valve for adjustment of the vacuum in the chamber
  • Replaceable valves with caps that allow individual vacuum control of each SPE column
  • Variable rack with exchangeable partitions, which accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials, etc.
Specifications
For Use With (Application) Solid Phase Extraction
For Use With (Equipment) up to 16 LV Columns
Type Vacuum Manifold
Quantity 1 Each