missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chromabond™ PTL Separation Column
Separate compounds with gravity flow phase separation. Chromabond™ PTL Separation Columns come in a variety of capacities
Brand: Macherey-Nagel™ 730738
Code : 50
Additional Details : Weight : 0.50000kg
Description
- Automatic separation of a two-phase mixture without separation funnel
- Two-phase mixtures are completely applied to the column and the phase boundary is determined without further work.
- The special membrane automatically stops the flow when the lower phase has passed.
- The upper phase remains in the column, thus both phases are available for futher analysis.
- Columns must not be run with vacuum or pressure
- PTL for solvents lighter than water, e.g., for diethyl ether, hexane
- maximum size 70 mL
Specifications
30 mL | |
For Solvents Lighter Than Water, Diethyl Ether, Hexane | |
Phase Separation Column |
SPE | |
Chromabond PTL |