Specifications
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 500/500 mg |
| For Use With (Application) | Solid Phase Extraction Analysis of PAH |
| Sorbent | Chromabond NH2/C18 |
| Type | Solid Phase Extraction Column |