Specifications
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 700/2000/700 mg |
| For Use With (Application) | Solid Phase Extraction Analysis of PCBs From Sludge |
| Sorbent | Chromabond NAN |
| Type | Solid Phase Extraction Column |