missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ Multi 96

Product Code. 12441768 Shop All Macherey-Nagel Products
Change view
Click to view available options
Material:
PTFE
Polypropylene
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
12441768 PTFE Each
12844511 Polypropylene Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12441768 Supplier Macherey-Nagel™ Supplier No. 738645
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Format Standard
Lid With Lid
Array 8 x 12
For Use With (Equipment) For Sealing of Individual Rows of The 96-Well Plate
Material PTFE
No. of Wells 96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.