Learn More
Macherey-Nagel™ Chromabond™ Multi 96
Product Code. 12844511 Shop All Macherey-Nagel Products
Change view
Click to view available options
Material:
PTFE
Polypropylene
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
12844511 Polypropylene Pack of 5
12441768 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12844511 Supplier Macherey-Nagel™ Supplier No. 738650.5
Specifications
Format Standard
Array 8 x 12
Material Polypropylene
No. of Wells 96
Total Volume (Metric) Well 96 x 2 mL
No. per Pack 5
Quantity 5/Pk.