Macherey Nagel™ Chromabond™ Easy Polypropylene SPE Column BIG Pack


Overview
Brand: Macherey Nagel™

Manufacturer Part Number: 730754.250

UNSPSC: 23151816

Code: 50

Additional Details:
Additional Details: Weight: 0.20000kg



Product Code. 12821031

Quantity Price
1 £ 840.0 / Each
Estimated Shipment
11-05-2017
Add to basket


Description and Specification

Specification

Pore Size 50Å
Column Capacity (Metric) 3mL
Column Type SPE
Particle Size 80μm
Product Type SPE Column
For Use With (Application) For SPE
Sorbent Chromabond Easy
Surface Area (Metric) 650 to 700m2/g