Specifications
| Base Material | Silica |
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| For Use With (Application) | Solid Phase Extraction |
| Sorbent | Chromabond Drug |
| Type | Solid Phase Extraction Column |