Learn More
Macherey-Nagel™ Chromabond™ Collecting Rack
Product Code. 10114661 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
12 Columns
16 LV Columns
24 Columns
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10114661 12 Columns Each
10368501 16 LV Columns Each
10072921 24 Columns Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10114661 Supplier Macherey-Nagel™ Supplier No. 730157
Only null left
Add to basket
Add to basket
Collection rack for use with SPE columns
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.
Specifications
For Use With (Application) Evaporation of Eluates
For Use With (Equipment) 12 Columns
Type Collecting Rack
Quantity 1 Each