missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ Alox N Polypropylene SPE Column

Brand:  Macherey-Nagel™ 730446

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10716132

  • £147.00 / Pack of 50
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

3 mL
500 mg
60 to 150 μm
Solid Phase Extraction Column
SPE
Solid Phase Extraction
Chromabond Alox N
150 m2/g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers