Specifications
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| For Use With (Application) | Solid Phase Extraction Analysis of Acrylamide in Food |
| Sorbent | Chromabond ABC18 |
| Type | Solid Phase Extraction Column |