missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHORDC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-78304
This item is not returnable.
View return policy
Description
CHORDC1 Polyclonal specifically detects CHORDC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CHORDC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| CHORD domain-containing protein 1, CHORD-containing protein 1, CHP1CHP-1, cysteine and histidine-rich domain (CHORD) containing 1, cysteine and histidine-rich domain (CHORD)-containing 1, cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1, cysteine and histidine-rich domain-containing protein 1, FLJ37289, Protein morgana | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.65 mg/mL | |
| Western Blot 1:2500, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunohistochemistry-Paraffin 1:150 | |
| Q9UHD1 | |
| CHORDC1 | |
| A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1] | |
| 0.1 mL | |
| Cancer, Epigenetics | |
| 26973 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction