missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHORDC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£173.00 - £362.00
Specifications
| Antigen | CHORDC1 |
|---|---|
| Concentration | 0.65 mg/mL |
| Dilution | Western Blot 1:2500, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunohistochemistry-Paraffin 1:150 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18098103
|
Novus Biologicals
NBP1-78304SS |
0.025 mL |
£173.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18290116
|
Novus Biologicals
NBP1-78304 |
0.1 mL |
£362.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHORDC1 Polyclonal specifically detects CHORDC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CHORDC1 | |
| Western Blot 1:2500, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunohistochemistry-Paraffin 1:150 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Epigenetics | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| CHORD domain-containing protein 1, CHORD-containing protein 1, CHP1CHP-1, cysteine and histidine-rich domain (CHORD) containing 1, cysteine and histidine-rich domain (CHORD)-containing 1, cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1, cysteine and histidine-rich domain-containing protein 1, FLJ37289, Protein morgana | |
| CHORDC1 | |
| IgG | |
| Affinity Purified |
| 0.65 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9UHD1 | |
| 26973 | |
| A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title