missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl acetate, 97+%, Thermo Scientific Chemicals

Product Code. 11407327
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407327 25 g 25g
11417327 100 g 100g
11427327 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407327 Supplier Thermo Scientific Alfa Aesar Supplier No. A15052.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cholesteryl acetate is used in cosmetics, wrist watches, thermometers, propane tank volume indicators, video displays, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl acetate is used in cosmetics, wrist watches, thermometers, propane tank volume indicators, video displays, pharmaceuticals.

Solubility
Soluble in chloroform, ethanol or ethyl acetate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-35-3
Molecular Formula C29H48O2
Molecular Weight (g/mol) 428.70
MDL Number MFCD00003636,MFCD00003636,MFCD00003636
InChI Key XUGISPSHIFXEHZ-UHFFFAOYNA-N
Synonym cholesteryl acetate
PubChem CID 12360660
SMILES CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(C)=O

Specifications

Melting Point 112°C to 114°C
Quantity 25 g
Beilstein 2064235
Merck Index 14,2201
Solubility Information Soluble in chloroform,ethanol or ethyl acetate.
Optical Rotation −43° (c=5 in Chloroform)
IUPAC Name [(10R,13R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Formula Weight 428.7
Percent Purity ≥97%
Chemical Name or Material Cholesteryl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.