CAS RN 604-35-3
CAS RN 604-35-3
IUPAC Name:
9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl acetate
Synonyms:
cholesteryl acetate
Molecular Weight (g/mol):
428.7
Molecular Formula:
C29H48O2
InChi Key:
XUGISPSHIFXEHZ-UHFFFAOYNA-N
SMILES:
CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(C)=O
1
–
1
of
1
results
1
Cholesteryl acetate, 97+%, Thermo Scientific Chemicals
CAS: 604-35-3 Molecular Formula: C29H48O2 Molecular Weight (g/mol): 428.70 MDL Number: MFCD00003636,MFCD00003636,MFCD00003636 InChI Key: XUGISPSHIFXEHZ-UHFFFAOYNA-N Synonym: cholesteryl acetate PubChem CID: 12360660 SMILES: CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(C)=O