missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl 2,4-dichlorobenzoate, Thermo Scientific™

Product Code. 11395447
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395447 50 g 50g
11305457 10 g 10g
11385447 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395447 Supplier Thermo Scientific Alfa Aesar Supplier No. 042700.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32832-01-2
Molecular Formula C34H48Cl2O2
Molecular Weight (g/mol) 559.656
MDL Number MFCD07776849
InChI Key NZZFKZMKJPWVDL-LPVBUYOKSA-N
Synonym cholesteryl-2,4-dichlor-benzoat, cholest-5-en-3??-yl 2,4-dichlorobenzoate, 3??-cholest-5-en-3-ol, 2,4-dichlorobenzoate
PubChem CID 122130190
IUPAC Name [(3S,9S,14S,17R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] 2,4-dichlorobenzoate
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C5=C(C=C(C=C5)Cl)Cl)C)C

Specifications

Melting Point 126°C
Quantity 50 g
Solubility Information Soluble in THF
Formula Weight 559.66
Physical Form Powder
Chemical Name or Material Cholesteryl 2,4-dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.