accessibility menu, dialog, popup

UserName

H3CCH3(R)CH3(R)(S)(S)(S)(R)H3C(R)H3COOClCl

CAS RN 32832-01-2

IUPAC Name: (1R,3aS,3bS,7S,9aR,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl 2,4-dichlorobenzoate
Synonyms: cholesteryl dichlorobenzoate
Molecular Weight (g/mol): 559.66
Molecular Formula: C34H48Cl2O2
InChi Key: NZZFKZMKJPWVDL-DTXYFFJBSA-N
SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)C3CC[C@]12C)OC(=O)C1=CC=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)