missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chocolate Agar with Vitox

Product Code. 13245359
Change view
Click to view available options
Description:
Chocolate Agar with Vitox
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Unit Size
13245359 Chocolate Agar with Vitox 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13245359 Supplier Thermo Scientific™ Supplier No. PO5090A
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and cultivate fastidious microorganisms with this highly nutritious medium which includes starch to absorb toxic metabolites produced by Neisseria.

Isolate and cultivate fastidious microorganisms using Thermo Scientific™ Chocolate Agar with Vitox, a highly nutritious medium which includes the presence of starch to ensure that toxic metabolites produced by Neisseria are absorbed.

This medium has been formulated to include a special peptone which is a mixture of meat and plant enzymatic digests for optimal performance and reliability.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Fawn brown, opaque
Content And Storage 2° to 12°C, in the dark
Description Chocolate Agar with Vitox
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.