missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55512
This item is not returnable.
View return policy
Description
CHMP1B Polyclonal specifically detects CHMP1B in Human samples. It is validated for Western Blot.
Specifications
| CHMP1B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C18orf2C10orf2, charged multivesicular body protein 1b, CHMP1.5C18-ORF2, CHMP1b, chromatin modifying protein 1B, Chromatin-modifying protein 1b, hVps46-2, vacuolar protein sorting 46-2, Vacuolar protein sorting-associated protein 46-2, Vps46-2, Vps46B | |
| Rabbit | |
| 22 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q7LBR1 | |
| CHMP1B | |
| Synthetic peptides corresponding to CHMP1B(chromatin modifying protein 1B) Antibody(against the N terminal of CHMP1B. Peptide sequence KIKKAIQKGNMEVARIHAENAIRQKNQAVNFLRMSARVDAVAARVQTAVT. | |
| Affinity purified | |
| RUO | |
| 57132 | |
| Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction