missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP1a Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CHMP1a |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668070
|
Novus Biologicals
NBP2-92043-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657511
|
Novus Biologicals
NBP2-92043-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHMP1a Polyclonal antibody specifically detects CHMP1a in Human, Mouse samples. It is validated for Western BlotSpecifications
| CHMP1a | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 5119 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CHMP1, CHMP1charged multivesicular body protein 1/chromatin modifying protein 1, chromatin modifying protein 1A, Chromatin-modifying protein 1a, hVps46-1, KIAA0047charged multivesicular body protein 1a, PCOLN3protease, metallo, 1, 33kD, procollagen (type III) N-endopeptidase, PRSM1CHMP1a, Vacuolar protein sorting-associated protein 46-1, VPS46-1, VPS46A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-196 of human CHMP1A (NP_002759.2). MDDTLFQLKFTAKQLEKLAKKAEKDSKAEQAKVKKALLQKNVECARVYAENAIRKKNEGVNWLRMASRVDAVASKVQTAVTMKGVTKNMAQVTKALDKALSTMDLQKVSSVMDRFEQQVQNLDVHTSVMEDSMSSATTLTTPQEQVDSLIMQIAEENGLEVLDQLSQLPEGASAVGESSVRSQEDQLSRRLAALRN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title