missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP1a Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92043-0.02ml
This item is not returnable.
View return policy
Description
CHMP1a Polyclonal antibody specifically detects CHMP1a in Human, Mouse samples. It is validated for Western Blot
Specifications
| CHMP1a | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CHMP1, CHMP1charged multivesicular body protein 1/chromatin modifying protein 1, chromatin modifying protein 1A, Chromatin-modifying protein 1a, hVps46-1, KIAA0047charged multivesicular body protein 1a, PCOLN3protease, metallo, 1, 33kD, procollagen (type III) N-endopeptidase, PRSM1CHMP1a, Vacuolar protein sorting-associated protein 46-1, VPS46-1, VPS46A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-196 of human CHMP1A (NP_002759.2). MDDTLFQLKFTAKQLEKLAKKAEKDSKAEQAKVKKALLQKNVECARVYAENAIRKKNEGVNWLRMASRVDAVASKVQTAVTMKGVTKNMAQVTKALDKALSTMDLQKVSSVMDRFEQQVQNLDVHTSVMEDSMSSATTLTTPQEQVDSLIMQIAEENGLEVLDQLSQLPEGASAVGESSVRSQEDQLSRRLAALRN | |
| 0.02 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| 5119 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction