missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine maleate, 99%, Thermo Scientific™

£25.50 - £62.00

Chemical Identifiers

CAS 113-92-8
Molecular Formula C20H23ClN2O4
Molecular Weight (g/mol) 390.86
MDL Number MFCD00069225
InChI Key DBAKFASWICGISY-BTJKTKAUNA-N
Synonym chlorpheniramine maleate, antagonate, allerclor, allergin, chlormene, alunex, carbinoxamide maleate, allergisan, -chlorpheniramine maleate salt, dl-2-p-chloro-a-2-dimethylamino ethylbenzyl pyridine bimaleate
PubChem CID 5281068
ChEBI CHEBI:3645
IUPAC Name [3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
SMILES OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11411000
SDS View Documents
Thermo Scientific Alfa Aesar
B25238.06
5g
£25.50
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11421000
SDS View Documents
Thermo Scientific Alfa Aesar
B25238.14
25g
£62.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

113-92-8
390.86
DBAKFASWICGISY-BTJKTKAUNA-N
5281068
[3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
C20H23ClN2O4
MFCD00069225
chlorpheniramine maleate, antagonate, allerclor, allergin, chlormene, alunex, carbinoxamide maleate, allergisan, -chlorpheniramine maleate salt, dl-2-p-chloro-a-2-dimethylamino ethylbenzyl pyridine bimaleate
CHEBI:3645
OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1

Specifications

113-92-8
MFCD00069225
DBAKFASWICGISY-BTJKTKAUNA-N
[3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
5281068
390.86
Chlorpheniramine maleate
C20H23ClN2O4
chlorpheniramine maleate, antagonate, allerclor, allergin, chlormene, alunex, carbinoxamide maleate, allergisan, -chlorpheniramine maleate salt, dl-2-p-chloro-a-2-dimethylamino ethylbenzyl pyridine bimaleate
OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
390.86
CHEBI:3645
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 204-037-5

missing translation for 'rtecsNumber' : US6503000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.