missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine maleate, 99%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar B25238.06

Quantity 5g

 View more versions of this product

Product Code. 11411000

  • £25.50 / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

113-92-8
390.86
DBAKFASWICGISY-BTJKTKAUNA-N
5281068
[3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
C20H23ClN2O4
MFCD00069225
chlorpheniramine maleate, antagonate, allerclor, allergin, chlormene, alunex, carbinoxamide maleate, allergisan, -chlorpheniramine maleate salt, dl-2-p-chloro-a-2-dimethylamino ethylbenzyl pyridine bimaleate
CHEBI:3645
OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1

Specifications

113-92-8
MFCD00069225
chlorpheniramine maleate, antagonate, allerclor, allergin, chlormene, alunex, carbinoxamide maleate, allergisan, -chlorpheniramine maleate salt, dl-2-p-chloro-a-2-dimethylamino ethylbenzyl pyridine bimaleate
OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
390.86
CHEBI:3645
99%
C20H23ClN2O4
5g
DBAKFASWICGISY-BTJKTKAUNA-N
[3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
5281068
390.86
Chlorpheniramine maleate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.