missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific Chemicals
£141.00 - £550.00
Chemical Identifiers
CAS | 58347-49-2 |
---|---|
Molecular Formula | C6H4ClN3 |
Molecular Weight (g/mol) | 153.57 |
MDL Number | MFCD04035684 |
InChI Key | YCZQHXPIKQHABJ-UHFFFAOYSA-N |
Synonym | 7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine |
PubChem CID | 12281647 |
IUPAC Name | 7-chloropyrazolo[1,5-a]pyrimidine |
SMILES | C1=C2N=CC=C(N2N=C1)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11314123
|
Thermo Scientific Acros
438490010 |
1 g | Glass bottle |
£141.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11344123
|
Thermo Scientific Acros
438490050 |
5 g | Glass bottle |
£550.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
58347-49-2 | |
153.57 | |
YCZQHXPIKQHABJ-UHFFFAOYSA-N | |
12281647 | |
C1=C2N=CC=C(N2N=C1)Cl |
C6H4ClN3 | |
MFCD04035684 | |
7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine | |
7-chloropyrazolo[1,5-a]pyrimidine |
Specifications
58347-49-2 | |
Brown to White | |
94% min. (GC) | |
MFCD04035684 | |
YCZQHXPIKQHABJ-UHFFFAOYSA-N | |
7-chloropyrazolo[1,5-a]pyrimidine | |
12281647 | |
95% | |
95% |
98.0°C to 102.0°C | |
Authentic | |
C6H4ClN3 | |
7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine | |
C1=C2N=CC=C(N2N=C1)Cl | |
153.57 | |
153.57 | |
Powder | |
7-Chloropyrazolo[1,5-a]pyrimidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 7-Chloropyrazolo¢1,5-a!pyrimidine, 95%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only