missing translation for 'onlineSavingsMsg'
Learn More

7-Chloropyrazolo[1,5-a]pyrimidine, 95%

Product Code. 11344123 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
missing translation for 'unitSize':
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
11344123 5 g Glass bottle 5g
11314123 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11344123 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 438490050
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 58347-49-2 | C6H4ClN3 | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58347-49-2
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.57
MDL Number MFCD04035684
InChI Key YCZQHXPIKQHABJ-UHFFFAOYSA-N
Synonym 7-chloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 7-chloro, 7-chloro-pyrazolo 1,5-a pyrimidine, zlchem 1087, 7-chloropyrazolo 1?5-a pyrimidine, 7-chloranylpyrazolo 1,5-a pyrimidine, 7-chloro-8-hydropyrazolo 1,5-a pyrimidine
PubChem CID 12281647
IUPAC Name 7-chloropyrazolo[1,5-a]pyrimidine
SMILES C1=C2N=CC=C(N2N=C1)Cl

Specifications

Melting Point 98.0°C to 102.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 153.57
Percent Purity 95%
Physical Form Powder
Assay 95%
Chemical Name or Material 7-Chloropyrazolo[1,5-a]pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 7-Chloropyrazolo[1,5-a]pyrimidine, 95%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.