missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red sodium salt, 0.04% w/v aq. soln., Thermo Scientific™

Product Code. 11358967
Change view
Click to view available options
Quantity:
500 mL
6 x 100 mL
Unit Size:
500mL
600mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358967 6 x 100 mL 600mL
11480151 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358967 Supplier Thermo Scientific Alfa Aesar Supplier No. 038699.M0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217203-49-0
MDL Number MFCD00151199

Specifications

Concentration 0.04% w/v aq. soln.
Formula Weight 445.26
Density 0.990 g/mL
Quantity 6 x 100 mL
Physical Form Liquid
Chemical Name or Material Chlorophenol Red sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.