CAS RN 1217203-49-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chlorophenol Red sodium salt, 0.04% w/v aq. soln., Thermo Scientific™
CAS: 1217203-49-0 MDL Number: MFCD00151199