missing translation for 'onlineSavingsMsg'
Learn More

chlorodimethylethylsilane, 97%, Thermo Scientific™

Product Code. 10341340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341340 5 g Glass bottle 5g
10301390 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341340 Supplier Thermo Scientific Acros Supplier No. 313500050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6917-76-6
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.67
MDL Number MFCD00009918
InChI Key AVDUEHWPPXIAEB-UHFFFAOYSA-N
Synonym ethyldimethylchlorosilane, chlorodimethylethylsilane, chloro ethyl dimethylsilane, dimethylethylchlorosilane, dimethylethylsilyl chloride, ethyldimethylsilyl chloride, chloro ethyl dimethylsilane #, silane, chlorodimethylethyl-,
PubChem CID 138863
IUPAC Name chloro-ethyl-dimethylsilane
SMILES CC[Si](C)(C)Cl

Specifications

Color Colorless
Density 0.8730g/mL
Boiling Point 89.0°C to 91.0°C
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.873
Formula Weight 122.67
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material chlorodimethylethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorodimethylethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.