accessibility menu, dialog, popup

UserName

H3CSiCH3CH3Cl

CAS RN 6917-76-6

IUPAC Name: chloro(ethyl)dimethylsilane
Synonyms: chloro(ethyl)dimethylsilane
Molecular Weight (g/mol): 122.67
Molecular Formula: C4H11ClSi
InChi Key: AVDUEHWPPXIAEB-UHFFFAOYSA-N
SMILES: CC[Si](C)(C)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results