missing translation for 'onlineSavingsMsg'
Learn More

Chloro(p-cymene)-N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I), 97%

Product Code. 15405135
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405135 1 g 1g
15415135 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405135 Supplier Thermo Scientific Alfa Aesar Supplier No. H30264.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 213603-12-4 | C23H33ClN2O2RuS

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213603-12-4
Molecular Formula C23H33ClN2O2RuS
MDL Number MFCD12546183
Synonym [(R,R)-TsDACH RuCl(p-cymene)]; (p-Cymene)N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I) chloride

Specifications

Quantity 1 g
Sensitivity Air sensitive
Formula Weight 538.11
Percent Purity 97%
Chemical Name or Material Chloro(p-cymene)-N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.