CAS RN 213603-12-4
CAS RN 213603-12-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chloro(p-cymene)-N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I), 97%
CAS: 213603-12-4 Molecular Formula: C23H33ClN2O2RuS MDL Number: MFCD12546183 Synonym: [(R,R)-TsDACH RuCl(p-cymene)]; (p-Cymene)N-(p-toluenesulfonyl)-(R,R)-1,2-cyclohexanediamineruthenium(I) chloride