Learn More
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Description
Cyclopentadienylbis(triphenylphosphine)ruthenium Chloride is used as an efficient transfer hydrogenation catalyst. It is used in the formation of indoles, isoquinolines and quinolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 135°C (decomposition) |
| Color | Orange |
| Quantity | 5 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in water. |
| IUPAC Name | Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct |
| Formula Weight | 772.27 |
| Physical Form | Crystalline Solid |
| Chemical Name or Material | Chloro (cyclopentadienyl)bis (triphenylphosphine)ruthenium (II) ethanol adduct |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.