CAS RN 32993-05-8
IUPAC Name:
chlororuthenium; cyclopentane; bis(triphenylphosphane)
Synonyms:
chlororuthenium; cyclopentane; bis(triphenylphosphine)
Molecular Weight (g/mol):
731.24
Molecular Formula:
C41H40ClP2Ru
InChi Key:
BJOHBGVDYMJYLL-UHFFFAOYSA-M
SMILES:
Cl[Ru].C1CCCC1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
CAS: 32993-05-8 Molecular Formula: C43H41ClOP2Ru Molecular Weight (g/mol): 772.27 MDL Number: MFCD00075004 InChI Key: YKZUFRITHQLOHK-UHFFFAOYSA-M Synonym: bis triphenylphosphine chloro cyclopentadienylruthenium,chlorocyclopentadienylbis triphenylphosphine ruthenium ii 095h PubChem CID: 132285030 SMILES: Cl[Ru++].CCO.c1cccc1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1