missing translation for 'onlineSavingsMsg'
Learn More

Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

Product Code. 15487205
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487205 1 g 1g
15497205 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487205 Supplier Thermo Scientific Alfa Aesar Supplier No. 046430.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32993-05-8 | C43H41ClOP2Ru | 772.27 g/mol

Cyclopentadienylbis(triphenylphosphine)ruthenium Chloride is used as an efficient transfer hydrogenation catalyst. It is used in the formation of indoles, isoquinolines and quinolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32993-05-8
Molecular Formula C43H41ClOP2Ru
Molecular Weight (g/mol) 772.27
MDL Number MFCD00075004
InChI Key YKZUFRITHQLOHK-UHFFFAOYSA-M
Synonym bis triphenylphosphine chloro cyclopentadienylruthenium, chlorocyclopentadienylbis triphenylphosphine ruthenium ii 095h
PubChem CID 132285030
SMILES Cl[Ru++].CCO.c1cccc1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 135°C (decomposition)
Color Orange
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water.
IUPAC Name chlororuthenium;cyclopentane;triphenylphosphane
Formula Weight 772.27
Physical Form Crystalline Solid
Chemical Name or Material Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.