missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Ebs Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
£166.00 - £400.00
Specifications
| Antigen | Chlamydia Ebs |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18043096
|
Novus Biologicals
NB100-64442-0.1ML |
0.1 mL |
£166.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18081524
|
Novus Biologicals
NB100-64442 |
0.5 mL |
£400.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlamydia Ebs Polyclonal specifically detects Chlamydia Ebs in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Chlamydia Ebs | |
| Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| IgG | |
| IgG purified | |
| Reacts with all antigens of strain L2 and purified elementary bodies of serovars A-K and L1-L3. Cross reactivity has been observed with C. psittacii and C. pneumoniae (TWAR). It does not react with HEp-2 cells or egg yolk sac proteins. |
| 1.0 mg/mL | |
| Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Goat | |
| Bacteria | |
| Chlamydia trachomatis, strain L2, and other serovar groups. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title