missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Ebs Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-64442
This item is not returnable.
View return policy
Description
Chlamydia Ebs Polyclonal specifically detects Chlamydia Ebs in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Chlamydia Ebs | |
| Polyclonal | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| Chlamydia trachomatis, strain L2, and other serovar groups. | |
| 0.5 mL | |
| Primary | |
| Bacteria | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Goat | |
| IgG purified | |
| RUO | |
| Reacts with all antigens of strain L2 and purified elementary bodies of serovars A-K and L1-L3. Cross reactivity has been observed with C. psittacii and C. pneumoniae (TWAR). It does not react with HEp-2 cells or egg yolk sac proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction