missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken RGM-A Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £346.00
Specifications
| Antigen | RGM-A |
|---|---|
| Clone | 292012 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18741039
![]() |
R&D Systems
MAB2010-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141911
|
R&D Systems
MAB2010 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGM-A Monoclonal specifically detects RGM-A in Chicken samples. It is validated for Western Blot.Specifications
| RGM-A | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chicken | |
| Q8JG54 | |
| 56963 | |
| Mouse myeloma cell line NS0-derived recombinant chicken RGM-A Pro150-Asn404 (Asn404Asp), predicted Accession # Q8JG54 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects chicken RGM-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human RGM-A or recombinant mouse RGM-A is observed. |
| 292012 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A | |
| RGMA | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title