missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken RGM-A Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2010
This item is not returnable.
View return policy
Description
RGM-A Monoclonal specifically detects RGM-A in Chicken samples. It is validated for Western Blot.
Specifications
| RGM-A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8JG54 | |
| RGMA | |
| Mouse myeloma cell line NS0-derived recombinant chicken RGM-A Pro150-Asn404 (Asn404Asp), predicted Accession # Q8JG54 | |
| 100 μg | |
| Primary | |
| Detects chicken RGM-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human RGM-A or recombinant mouse RGM-A is observed. | |
| Chicken | |
| Purified |
| Western Blot | |
| 292012 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56963 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction