Learn More
3M™ Chemical Protective Coverall 4570
Product Code. p-200137731 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
31297040 Small
31297036 Medium
31297037 Large
31297038 X-Large
31297039 2X-Large
31297034 3X-Large
31297035 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 31297040 Supplier 3M™ Supplier No. 7100256137
Add to basket
Add to basket
Chemical protective suit, certified to Type 3/4/5/6 levels of protection to help protect against a range of chemical jets and sprays.
  • Made from strong 92 GSM non-microporous PE laminate for durability and protection.
  • Double front closing system and large zip pull ensure easy wear and secure fit.
  • Provides excellent barrier protection against liquid splashes, jet sprays (Type 3), and hazardous particles (TIL Class 2, EN 1073-2).
  • Meets multiple standards: EN 14126, EN 1073-2, EN 1149, EN ISO 13688, Type 6 (EN 13034), Type 5 (EN ISO 13982-1), Type 4 and Type 3 (EN 14605).
  • Gray color design for low-visibility and discreet applications such as first responders.
  • Chemically resistant taped seams provide enhanced protection and strength.
  • High chemical permeation resistance tested against multiple chemicals.
  • Passes all EN 14126 tests, achieving the highest protection class.
  • Offers high particle holdout (TIL Class 2, EN 1073-2).
  • Reliable safety and protection for the workforce.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Small
Gender Unisex
Material Polyethylene, Polypropylene
Color Gray
Disposable Single-use
Waist Style Standard
Ankle Style Open
Cuff Style Open
Neck Style Hooded
Hood Type Open Face
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Chemicals, Non-toxic Particulates