missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chemerin Monoclonal Antibody (D2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550909
This item is not returnable.
View return policy
Description
Retinoids act through ligand-dependent transcription factors, retinoid X receptor (RXRs) and retinoic acid receptors (RARs). Tazarotene-induced gene (TIG) proteins, also designated retinoic acid receptor responder proteins or RAR-responsive proteins, can be membrane bound or secreted. TIGs act as tumor suppressor genes in human cancers and are highly expressed in skin, hair follicles and endothelial cells as well as in pancreas, spleen and intestine. TIGs are activated by tazarotene and have been implicated as growth regulators that mediate the growth suppressive effects of retinoids. TIG2 is a secreted protein that is mainly expressed in epidermis, hair follicles and endothelial cells. TIG2 is inhibited in psoriatic lesions and is activated by tazarotene in skin rafts and in epidermis of psoriatic lesions.
Specifications
| Chemerin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 0610007L05Rik; AI303516; Chemerin; HP10433; Rarres2; RAR-responsive protein TIG2; retinoic acid receptor responder (tazarotene induced) 2; retinoic acid receptor responder 2; retinoic acid receptor responder protein 2; Tazarotene-induced gene 2 protein; TIG2; TIG-2; TIG2HP10433 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 297073 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| D2 | |
| Unconjugated | |
| Rarres2 | |
| Rarres2 | |
| Recombinant Rat CHEM protein, Ser25-Gly152 (Accession #Q5BK77). | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction