missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Chelating Sepharose™ Fast Flow Medium

Product Code. 10677085 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10677085 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10677085 Supplier Cytiva Supplier No. 17057501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for purifying native and recombinant proteins and peptides that have an affinity for metal ions. Chelating Sepharose™ Fast Flow Medium provides robust performance and regulatory support.

  • Immobilised Metal Ion Affinity chromatography media fulfills industrial demands for security of supply, robust performance and regulatory support
  • Supplied free of metal ions allowing possibility to charge with your metal ion of choice for optimized selectivity
  • Cross-linked 6% agarose beads modified with iminodiacetic immobilized to the base matrix by stable ether linkages and sufficiently long spacer arms

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 20% Ethanol, 8M Guanidine Hydrochloride, 8M Urea, 0.01M HCl, non-ionic Detergent, Acetate Buffer pH 4
Product Type Chelating Sepharose FF medium
Quantity 50 mL
Height (English) 9.84 in.
Height (Metric) 250mm
Ligand Type Iminodiacetic Acid
Max. Pressure 10psi
pH Range 3 to 13
Particle Size 90μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) Purifying native and recombinant Proteins and Peptides that have an affinity for Metal ions
Matrix Chelating Sepharose
Show More Show Less
compliance-icons
Product Identifier
  • Chelating Sepharose Fast Flow, 50 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.