missing translation for 'onlineSavingsMsg'
Learn More

CHCHD2 Mouse anti-Human, Porcine, Rat, Clone: 2G1G10, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen CHCHD2
Clone 2G1G10
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16838055
View Documents
Proteintech
66302-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16828055
View Documents
Proteintech
66302-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).
Specifications

Specifications

CHCHD2
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
C7orf17, CHCHD2, NS2TP
Chchd2
IgG1
Protein G
Antibody
2G1G10
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Pig, Rat
Q9Y6H1
100518545, 316643, 51142
CHCHD2 Fusion Protein Ag24219
Primary
-20°C
CHCHD2
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.